2-Bromo-α-ethenyl-6-fluorobenzenemethanol ≥96%

Supplier: AOBChem USA

Warning

235088-1G 235088-500MG 235088-5G
77467-188EA 654.05 USD
77467-188 77479-912 77490-218
2-Bromo-α-ethenyl-6-fluorobenzenemethanol ≥96%
2-Bromo-α-ethenyl-6-fluorobenzenemethanol

Precaución: For research use only

Formula: C₉H₈BrFO
Storage Temperature: Ambient
MDL Number: MFCD26138839
CAS Number: 1378432-96-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR