2-Chloro-α-ethenyl-6-fluorobenzenemethanol ≥97%

Supplier: AOBChem USA

Warning

235079-1G 235079-500MG 235079-5G
77467-184EA 751.9 USD
77467-184 77479-908 77490-214
2-Chloro-α-ethenyl-6-fluorobenzenemethanol ≥97%
2-Chloro-α-ethenyl-6-fluorobenzenemethanol

Precaución: For research use only

Formula: C₉H₈ClFO
MW: 186.61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21232099
CAS Number: 149946-87-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR