α-Ethenyl-3-(methylthio)benzenemethanol ≥97%

Supplier: AOBChem USA

Warning

234968-1G 234968-500MG 234968-5G
77467-180EA 751.9 USD
77467-180 77479-904 77490-208
α-Ethenyl-3-(methylthio)benzenemethanol ≥97%
α-Ethenyl-3-(methylthio)benzenemethanol

Precaución: For research use only

Formula: C₁₀H₁₂OS
MW: 180.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34870203
CAS Number: 2141366-00-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR