α-Ethenyl-2-(1-methylethyl)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

234963-1G 234963-500MG 234963-5G
77467-176EA 553.11 USD
77467-176 77479-900 77490-204
α-Ethenyl-2-(1-methylethyl)benzenemethanol ≥95%
α-Ethenyl-2-(1-methylethyl)benzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₆O
MW: 176.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34870200
CAS Number: 2229387-70-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR