α-Ethenyl-2,5-dimethylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

234942-1G 234942-500MG 234942-5G 234942-25G
77467-172EA 380.07 USD
77467-172 77479-896 77490-200 77500-398
α-Ethenyl-2,5-dimethylbenzenemethanol ≥95%
α-Ethenyl-2,5-dimethylbenzenemethanol

Precaución: For research use only

Formula: C₁₁H₁₄O
Storage Temperature: Ambient
MDL Number: MFCD21232125
CAS Number: 945030-90-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR