α-Ethenyl-4-ethylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

234919-1G 234919-500MG 234919-5G 234919-25G
77467-168EA 380.07 USD
77467-168 77479-892 77490-196 77500-394
α-Ethenyl-4-ethylbenzenemethanol ≥95%
α-Ethenyl-4-ethylbenzenemethanol

Precaución: For research use only

Formula: C₁₁H₁₄O
Storage Temperature: Ambient
MDL Number: MFCD21232115
CAS Number: 1365540-83-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR