α-Ethenyl-2-fluorobenzenemethanol ≥96%

Supplier: AOBChem USA

Warning

234912-1G 234912-500MG 234912-5G
77467-164EA 810.61 USD
77467-164 77479-888 77490-192
α-Ethenyl-2-fluorobenzenemethanol ≥96%
α-Ethenyl-2-fluorobenzenemethanol

Precaución: For research use only

Formula: C₉H₉FO
MW: 152.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21232107
CAS Number: 58824-51-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR