α-Ethenyl-4-fluorobenzenemethanol ≥96%

Supplier: AOBChem USA

Warning

234902-1G 234902-500MG 234902-5G
77467-160EA 715.85 USD
77467-160 77479-884 77490-188
α-Ethenyl-4-fluorobenzenemethanol ≥96%
α-Ethenyl-4-fluorobenzenemethanol

Precaución: For research use only

Formula: C₉H₉FO
MW: 152.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21232100
CAS Number: 5724-03-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR