3-Bromo-2,6-difluoro-α-methyl-α-phenylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

234843-1G 234843-500MG 234843-5G 234843-25G
77467-156EA 415.09 USD
77467-156 77479-880 77490-184 77500-388
3-Bromo-2,6-difluoro-α-methyl-α-phenylbenzenemethanol ≥95%
3-Bromo-2,6-difluoro-α-methyl-α-phenylbenzenemethanol

Precaución: For research use only

Formula: C₁₄H₁₁BrF₂O
Storage Temperature: Ambient
MDL Number: MFCD34870181
CAS Number: 1991802-83-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR