α-Methyl-α-phenyl[1,1'-biphenyl]-4-methanol ≥95%

Supplier: AOBChem USA

Warning

234765-1G 234765-500MG 234765-5G 234765-25G
77467-152EA 415.09 USD
77467-152 77479-876 77490-180 77500-384
α-Methyl-α-phenyl[1,1'-biphenyl]-4-methanol ≥95%
α-Methyl-α-phenyl[1,1'-biphenyl]-4-methanol

Precaución: For research use only

Formula: C₂₀H₁₈O
MW: 274.36 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34870153
CAS Number: 622408-02-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR