1-(2,4-dimethoxyphenyl)ethanol ≥95%

Supplier: AOBChem USA

Warning

234747-1G 234747-500MG 234747-5G 234747-25G
77467-148EA 341.96 USD
77467-148 77479-872 77490-176 77500-382
1-(2,4-dimethoxyphenyl)ethanol ≥95%
1-(2,4-dimethoxyphenyl)ethanol

Precaución: For research use only

Formula: C₁₀H₁₄O₃
MW: 182.2163 g/mol
Storage Temperature: Ambient
CAS Number: 829-19-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR