1-(6-Methoxy-2-pyridyl)piperazine ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 1-(6-Methoxy-2-pyridinyl)piperazine

23467-1G 23467-500MG 23467-5G 23467-25G
77467-144EA 96.82 USD
77467-144 77479-868 77490-172 77500-378
1-(6-Methoxy-2-pyridyl)piperazine ≥95%
1-(6-Methoxy-2-pyridyl)piperazine

Precaución: For research use only

Formula: C₁₀H₁₅N₃O
Storage Temperature: Ambient
MDL Number: MFCD06738851
CAS Number: 51047-54-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR