Cyclopentyl(3,5-dimethoxyphenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

234554-1G 234554-500MG 234554-5G
77467-140EA 576.8 USD
77467-140 77479-864 77490-168
Cyclopentyl(3,5-dimethoxyphenyl)methanone ≥95%
Cyclopentyl(3,5-dimethoxyphenyl)methanone

Precaución: For research use only

Formula: C₁₄H₁₈O₃
MW: 234.29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24796797
CAS Number: 623559-88-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR