1-(3,5-Dimethoxyphenyl)-2-methyl-1-propanone ≥96%

Supplier: AOBChem USA

Warning

234455-1G 234455-500MG 234455-5G
77467-136EA 576.8 USD
77467-136 77479-860 77490-164
1-(3,5-Dimethoxyphenyl)-2-methyl-1-propanone ≥96%
1-(3,5-Dimethoxyphenyl)-2-methyl-1-propanone

Precaución: For research use only

Formula: C₁₂H₁₆O₃
MW: 208.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20920735
CAS Number: 73109-77-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR