Methyl 4-fluoro-3-Methyl-α-oxobenzeneacetate ≥95%

Supplier: AOBChem USA

Warning

234386-1G 234386-500MG 234386-5G
77467-132EA 648.9 USD
77467-132 77479-856 77490-160
Methyl 4-fluoro-3-Methyl-α-oxobenzeneacetate ≥95%
Methyl 4-fluoro-3-Methyl-α-oxobenzeneacetate

Precaución: For research use only

Formula: C₁₀H₉FO₃
MW: 196.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11617600
CAS Number: 1267230-81-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR