1-(3,5-Dimethylphenyl)-2-methyl-1-propanone ≥96%

Supplier: AOBChem USA

Warning

234189-1G 234189-500MG 234189-5G 234189-25G
77467-128EA 333.72 USD
77467-128 77479-852 77490-156 77500-376
1-(3,5-Dimethylphenyl)-2-methyl-1-propanone ≥96%
1-(3,5-Dimethylphenyl)-2-methyl-1-propanone

Precaución: For research use only

Formula: C₁₂H₁₆O
MW: 176.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12806383
CAS Number: 827348-35-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR