4-Chloro-2,3-difluoro-5-hydroxybenzoic acid ≥95%

Supplier: AOBChem USA

Warning

23407-1G 23407-500MG 23407-5G
77467-120EA 526.33 USD
77467-120 77479-844 77490-148
4-Chloro-2,3-difluoro-5-hydroxybenzoic acid ≥95%
4-Chloro-2,3-difluoro-5-hydroxybenzoic acid

Precaución: For research use only

Formula: C₇H₃ClF₂O₃
MW: 208.547 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22056766
CAS Number: 749230-50-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR