1-(3,5-Dimethylphenyl)-1-butanone ≥95%

Supplier: AOBChem USA

Warning

234040-1G 234040-500MG 234040-5G 234040-25G
77467-116EA 300.76 USD
77467-116 77479-840 77490-144 77500-368
1-(3,5-Dimethylphenyl)-1-butanone ≥95%
1-(3,5-Dimethylphenyl)-1-butanone

Precaución: For research use only

Formula: C₁₂H₁₆O
Storage Temperature: Ambient
MDL Number: MFCD11617493
CAS Number: 61088-39-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR