1-Butanone, 1-(3-fluoro-4-methylphenyl)- ≥95%

Supplier: AOBChem USA

Warning

233605-1G 233605-500MG 233605-5G 233605-25G
77467-108EA 300.76 USD
77467-108 77479-832 77490-136 77500-362
1-Butanone, 1-(3-fluoro-4-methylphenyl)- ≥95%
1-Butanone, 1-(3-fluoro-4-methylphenyl)-

Precaución: For research use only

Formula: C₁₁H₁₃FO
MW: 180.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11617475
CAS Number: 1178929-83-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR