1-(3-Fluoro-4-methylphenyl)-1-propanone ≥95%

Supplier: AOBChem USA

Warning

233258-1G 233258-500MG 233258-5G 233258-25G
77467-104EA 287.37 USD
77467-104 77479-828 77490-132 77500-358
1-(3-Fluoro-4-methylphenyl)-1-propanone ≥95%
1-(3-Fluoro-4-methylphenyl)-1-propanone

Precaución: For research use only

Formula: C₁₀H₁₁FO
MW: 166.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11617649
CAS Number: 107076-07-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR