1-(4-Bromo-6-fluoro-1H-indazol-1-yl)ethanone ≥95%

Supplier: AOBChem USA

Warning

23274-1G 23274-500MG 23274-5G 23274-25G
77467-100EA 269.86 USD
77467-100 77479-824 77490-128 77500-354
1-(4-Bromo-6-fluoro-1H-indazol-1-yl)ethanone ≥95%
1-(4-Bromo-6-fluoro-1H-indazol-1-yl)ethanone

Precaución: For research use only

Formula: C₉H₆BrFN₂O
MW: 257.059 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18909370
CAS Number: 1333222-13-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR