2,6-Dimethoxy-4-fluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

23141-1G 23141-500MG 23141-5G
77467-096EA 540.75 USD
77467-096 77479-820 77490-124
2,6-Dimethoxy-4-fluorobenzaldehyde ≥97%
2,6-Dimethoxy-4-fluorobenzaldehyde

Precaución: For research use only

Formula: C₉H₉FO₃
MW: 184.164 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12025137
CAS Number: 139549-11-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR