1-(Naphthalen-2-yl)propan-1-one ≥96%

Supplier: AOBChem USA

Warning

230338-1G 230338-500MG 230338-5G 230338-25G
77467-092EA 160.68 USD
77467-092 77479-816 77490-120 77500-350
1-(Naphthalen-2-yl)propan-1-one ≥96%
1-(Naphthalen-2-yl)propan-1-one

Precaución: For research use only

Formula: C₁₃H₁₂O
MW: 184.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00228290
CAS Number: 6315-96-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR