1-Butyl-4-[(1-methylethyl)thio]benzene ≥95%

Supplier: AOBChem USA

Warning

229429-1G 229429-500MG 229429-5G 229429-25G
77467-080EA 415.09 USD
77467-080 77479-804 77490-106 77500-340
1-Butyl-4-[(1-methylethyl)thio]benzene ≥95%
1-Butyl-4-[(1-methylethyl)thio]benzene

Precaución: For research use only

Formula: C₁₃H₂₀S
MW: 208.36 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31621780
CAS Number: 85109-93-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR