1-(1-Methylethyl)-3-[(1-methylethyl)thio]benzene ≥95%

Supplier: AOBChem USA

Warning

229404-1G 229404-500MG 229404-5G 229404-25G
77467-076EA 415.09 USD
77467-076 77479-800 77490-102 77500-336
1-(1-Methylethyl)-3-[(1-methylethyl)thio]benzene ≥95%
1-(1-Methylethyl)-3-[(1-methylethyl)thio]benzene

Precaución: For research use only

Formula: C₁₂H₁₈S
MW: 194.34 g/mol
Storage Temperature: Ambient
CAS Number: 2254574-69-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR