1-[(1-Methylethyl)thio]-3-(methylthio)benzene ≥95%

Supplier: AOBChem USA

Warning

229393-1G 229393-500MG 229393-5G 229393-25G
77467-072EA 415.09 USD
77467-072 77479-796 77490-098 77500-332
1-[(1-Methylethyl)thio]-3-(methylthio)benzene ≥95%
1-[(1-Methylethyl)thio]-3-(methylthio)benzene

Precaución: For research use only

Formula: C₁₀H₁₄S₂
MW: 198.35 g/mol
Storage Temperature: Ambient
CAS Number: 79128-11-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR