1-(1,1-Dimethylethyl)-3-[(1-methylethyl)thio]benzene ≥95%

Supplier: AOBChem USA

Warning

229362-1G 229362-500MG 229362-5G 229362-25G
77467-068EA 415.09 USD
77467-068 77479-792 77490-094 77500-328
1-(1,1-Dimethylethyl)-3-[(1-methylethyl)thio]benzene ≥95%
1-(1,1-Dimethylethyl)-3-[(1-methylethyl)thio]benzene

Precaución: For research use only

Formula: C₁₃H₂₀S
MW: 208.36 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31621816
CAS Number: 2254576-13-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR