1-Ethoxy-3-[(1-methylethyl)thio]benzene ≥95%

Supplier: AOBChem USA

Warning

229352-1G 229352-500MG 229352-5G 229352-25G
77467-064EA 415.09 USD
77467-064 77479-788 77490-090 77500-324
1-Ethoxy-3-[(1-methylethyl)thio]benzene ≥95%
1-Ethoxy-3-[(1-methylethyl)thio]benzene

Precaución: For research use only

Formula: C₁₁H₁₆OS
MW: 196.31 g/mol
Storage Temperature: Ambient
CAS Number: 2254577-91-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR