1-Methyl-2-[(1-methylethyl)thio]benzene ≥95%
Supplier: AOBChem USA
|
Warning
|
229314-1G
229314-500MG
229314-5G
77467-060EA
696.28
USD
77467-060
77479-784
77490-086
1-Methyl-2-[(1-methylethyl)thio]benzene ≥95%
1-Methyl-2-[(1-methylethyl)thio]benzene
Precaución: For research use only
Formula:
C₁₀H₁₄S MW: 166.28 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD31621835 CAS Number: 15560-98-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...