tert-Butyl 3-methylbenzoate ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 1,1-Dimethylethyl 3-methylbenzoate

229109-1G 229109-500MG 229109-5G 229109-25G
77467-056EA 96.82 USD
77467-056 77479-780 77490-082 77500-320
tert-Butyl 3-methylbenzoate ≥95%
tert-Butyl 3-methylbenzoate

Precaución: For research use only

Formula: C₁₂H₁₆O₂
MW: 192.254 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16620897
CAS Number: 16537-19-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR