4-Bromo-3-fluoro-2-methoxypyridine ≥95%

Supplier: AOBChem USA

Warning

22656-1G 22656-500MG 22656-5G
77467-048EA 727.18 USD
77467-048 77479-772 77490-072
4-Bromo-3-fluoro-2-methoxypyridine ≥95%
4-Bromo-3-fluoro-2-methoxypyridine

Precaución: For research use only

Formula: C₆H₅BrFNO
Storage Temperature: Ambient
MDL Number: MFCD16609960
CAS Number: 1227599-41-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR