2-Chloro-1-ethyl-4-nitrobenzene ≥97%

Supplier: AOBChem USA

Warning

226249-1G 226249-5G 226249-25G
77467-040EA 60.77 USD
77467-040 77490-064 77500-306
2-Chloro-1-ethyl-4-nitrobenzene ≥97%
2-Chloro-1-ethyl-4-nitrobenzene

Precaución: For research use only

Formula: C₈H₈ClNO₂
Storage Temperature: Ambient
MDL Number: MFCD00034529
CAS Number: 42782-54-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR