N-(4-Fluoro-2-(trifluoromethyl)phenyl)acetamide ≥97%

Supplier: AOBChem USA

Warning

226118-1G 226118-500MG 226118-5G 226118-25G
77467-036EA 368.74 USD
77467-036 77479-762 77490-060 77500-302
N-(4-Fluoro-2-(trifluoromethyl)phenyl)acetamide ≥97%
N-(4-Fluoro-2-(trifluoromethyl)phenyl)acetamide

Precaución: For research use only

Formula: C₉H₇F₄NO
MW: 221.1515 g/mol
Storage Temperature: Ambient
MDL Number: MFCD25963078
CAS Number: 393-23-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR