2-(4-Chlorophenoxy)pyridine-5-boronic acid ≥97%

Supplier: AOBChem USA

Warning

22439-1G 22439-500MG 22439-5G
77467-028EA 653.02 USD
77467-028 77479-754 77490-052
2-(4-Chlorophenoxy)pyridine-5-boronic acid ≥97%
2-(4-Chlorophenoxy)pyridine-5-boronic acid

Precaución: For research use only

Formula: C₁₁H₉BClNO₃
MW: 249.458 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12403524
CAS Number: 1311183-19-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR