2,6-Dichloro-4-(methylsulfonyl)benzoic acid ≥96%

Supplier: AOBChem USA

Warning

223392-1G 223392-500MG 223392-5G 223392-25G
77467-020EA 408.91 USD
77467-020 77479-746 77490-044 77500-286
2,6-Dichloro-4-(methylsulfonyl)benzoic acid ≥96%
2,6-Dichloro-4-(methylsulfonyl)benzoic acid

Precaución: For research use only

Formula: C₈H₆Cl₂O₄S
MW: 269.1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09999670
CAS Number: 1121585-09-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR