4-Chloro-3-(methylsulfonyl)benzoic acid ≥96%

Supplier: AOBChem USA

Warning

223332-1G 223332-500MG 223332-5G 223332-25G
77467-016EA 258.53 USD
77467-016 77479-742 77490-040 77500-282
4-Chloro-3-(methylsulfonyl)benzoic acid ≥96%
4-Chloro-3-(methylsulfonyl)benzoic acid

Precaución: For research use only

Formula: C₈H₇ClO₄S
MW: 234.66 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06342742
CAS Number: 51522-07-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR