1,4-Difluoro-2-(methylsulfonyl)benzene ≥96%

Supplier: AOBChem USA

Warning

223129-1G 223129-5G 223129-25G
77467-012EA 60.77 USD
77467-012 77490-034 77500-278
1,4-Difluoro-2-(methylsulfonyl)benzene ≥96%
1,4-Difluoro-2-(methylsulfonyl)benzene

Precaución: For research use only

Formula: C₇H₆F₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD06800566
CAS Number: 236739-03-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR