2-Fluoro-6-methylpyridine-3-carboxaldehyde ≥97%

Supplier: AOBChem USA

Warning

22222-1G 22222-500MG 22222-5G 22222-25G
77467-004EA 261.62 USD
77467-004 77479-732 77490-026 77500-270
2-Fluoro-6-methylpyridine-3-carboxaldehyde ≥97%
2-Fluoro-6-methylpyridine-3-carboxaldehyde

Precaución: For research use only

Formula: C₇H₆FNO
MW: 139.127 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16611275
CAS Number: 1227564-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR