4-Fluoro-2-isopropoxyphenol ≥95%

Supplier: AOBChem USA

Warning

22210-1G 22210-500MG 22210-5G 22210-25G
77467-000EA 391.4 USD
77467-000 77479-728 77490-022 77500-268
4-Fluoro-2-isopropoxyphenol ≥95%
4-Fluoro-2-isopropoxyphenol

Precaución: For research use only

Formula: C₉H₁₁FO₂
MW: 170.181 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16997296
CAS Number: 1243454-07-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR