2,4-Dimethoxypyridin-3-ylboronic acid ≥95%

Supplier: AOBChem USA

Warning

22136-1G 22136-500MG 22136-5G
77466-996EA 582.98 USD
77466-996 77479-724 77490-018
2,4-Dimethoxypyridin-3-ylboronic acid ≥95%
2,4-Dimethoxypyridin-3-ylboronic acid

Precaución: For research use only

Formula: C₇H₁₀BNO₄
Storage Temperature: Ambient
MDL Number: MFCD18255780
CAS Number: 1219744-53-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR