2-Bromo-1-(3-Bromo-4-chlorophenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 2-Bromo-1-(3-bromo-4-chlorophenyl)ethanone

220906-1G 220906-500MG 220906-5G
77466-992EA 515 USD
77466-992 77479-720 77490-014
2-Bromo-1-(3-Bromo-4-chlorophenyl)ethanone ≥95%
2-Bromo-1-(3-Bromo-4-chlorophenyl)ethanone

Precaución: For research use only

Formula: C₈H₅Br₂ClO
MW: 312.38 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18392526
CAS Number: 87427-58-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR