2-Propanoyl-3-picoline ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 1-(3-Methylpyridin-2-yl)propan-1-one

220215-1G 220215-500MG 220215-5G 220215-25G
77466-984EA 93.73 USD
77466-984 77479-712 77490-006 77500-256
2-Propanoyl-3-picoline ≥95%
2-Propanoyl-3-picoline

Precaución: For research use only

Formula: C₉H₁₁NO
MW: 149.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16087790
CAS Number: 917872-34-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR