1-(6-Bromo-2-chloro-3-pyridinyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

220075-1G 220075-500MG 220075-5G
77466-980EA 487.19 USD
77466-980 77479-708 77490-000
1-(6-Bromo-2-chloro-3-pyridinyl)ethanone ≥95%
1-(6-Bromo-2-chloro-3-pyridinyl)ethanone

Precaución: For research use only

Formula: C₇H₅BrClNO
MW: 234.48 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18255735
CAS Number: 1256836-49-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR