1-(Methylsulfonyl)-4-(trifluoromethyl)benzene ≥96%

Supplier: AOBChem USA

Warning

219595-1G 219595-500MG 219595-5G 219595-25G
77466-976EA 317.24 USD
77466-976 77479-704 77489-996 77500-246
1-(Methylsulfonyl)-4-(trifluoromethyl)benzene ≥96%
1-(Methylsulfonyl)-4-(trifluoromethyl)benzene

Precaución: For research use only

Formula: C₈H₇F₃O₂S
MW: 224.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03789180
CAS Number: 145963-48-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR