3-Ethoxy-5-(trifluoromethyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

21947-1G 21947-500MG 21947-5G
77466-972EA 541.78 USD
77466-972 77479-700 77489-992
3-Ethoxy-5-(trifluoromethyl)benzaldehyde ≥97%
3-Ethoxy-5-(trifluoromethyl)benzaldehyde

Precaución: For research use only

Formula: C₁₀H₉F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD19105394
CAS Number: 1310416-64-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR