1-(3,5-Dibromo-2-fluorophenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

219177-1G 219177-500MG 219177-5G
77466-968EA 485.13 USD
77466-968 77479-696 77489-986
1-(3,5-Dibromo-2-fluorophenyl)ethanone ≥95%
1-(3,5-Dibromo-2-fluorophenyl)ethanone

Precaución: For research use only

Formula: C₈H₅Br₂FO
Storage Temperature: Ambient
MDL Number: MFCD28784563
CAS Number: 1807055-66-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR