2,2,2-Trifluoro-1-(2-fluoropyridin-3-yl)ethanone ≥97%

Supplier: AOBChem USA

Warning

21908-1G 21908-500MG 21908-5G 21908-25G
77466-964EA 168.92 USD
77466-964 77479-692 77489-982 77500-236
2,2,2-Trifluoro-1-(2-fluoropyridin-3-yl)ethanone ≥97%
2,2,2-Trifluoro-1-(2-fluoropyridin-3-yl)ethanone

Precaución: For research use only

Formula: C₇H₃F₄NO
Storage Temperature: Refrigerator
MDL Number: MFCD11847265
CAS Number: 1186608-79-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR