3,6-Dimethoxy-4-pyridazinecarboxaldehyde ≥97%

Supplier: AOBChem USA

Warning

21905-1G 21905-500MG 21905-5G
77466-960EA 616.97 USD
77466-960 77479-688 77489-978
3,6-Dimethoxy-4-pyridazinecarboxaldehyde ≥97%
3,6-Dimethoxy-4-pyridazinecarboxaldehyde

Precaución: For research use only

Formula: C₇H₈N₂O₃
Storage Temperature: Ambient
MDL Number: MFCD24684087
CAS Number: 262353-18-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR