5-Bromo-2,3,4-trifluorophenol ≥95%

Supplier: AOBChem USA

Warning

21860-1G 21860-500MG 21860-5G 21860-25G
77466-956EA 195.7 USD
77466-956 77479-684 77489-974 77500-230
5-Bromo-2,3,4-trifluorophenol ≥95%
5-Bromo-2,3,4-trifluorophenol

Precaución: For research use only

Formula: C₆H₂BrF₃O
MW: 226.979 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12405433
CAS Number: 1073339-19-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR