2-Bromo-1-chloro-4-fluoro-3-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

21803-1G 21803-500MG 21803-5G 21803-25G
77466-952EA 420.24 USD
77466-952 77479-680 77489-970 77500-226
2-Bromo-1-chloro-4-fluoro-3-methylbenzene ≥97%
2-Bromo-1-chloro-4-fluoro-3-methylbenzene

Precaución: For research use only

Formula: C₇H₅BrClF
MW: 223.47 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28141610
CAS Number: 1806971-78-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR